Learn More
Sigma Aldrich Fine Chemicals Biosciences ReadyScriptR cDNA Synthesi
Catalog No. 502914688
Encompass
ReadyScript cDNA Synthesis Mix is a sensitive and easy-to-use SOLUTIONon for two-step RT-PCR. This 5X concentrated mix provides all necessary components (except RNA template) for first-strand synthesis including buffer dNTPs MgCl2 primers RNase inhibitor protein ReadyScript reverse transcriptase and stabilizers. ReadyScript is a RNase H() derivative of MMLV reverse transcriptase optimized for reliable cDNA synthesis over a wide dynamic range of input RNA. The unique blend of oligo (dT) and random primers in the ReadyScript cDNA Synthesis Mix works exceptionally well with a wide variety of targets. It is optimized for the production of targets 1kb in length. ReadyScript cDNA Synthesis Mix produces excellent results in both real-time and conventional RT-PCR.
Catalog No. 50-291-4688 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RDRT500RXN
ReadyScript cDNA Synthesis Mix is a sensitive and easy-to-use SOLUTIONon for two-step RT-PCR. This 5X concentrated mix provides all necessary components (except RNA template) for first-strand synthesis including buffer dNTPs MgCl2 primers RNase inhibitor protein ReadyScript reverse transcriptase and stabilizers. ReadyScript is a RNase H() derivative of MMLV reverse transcriptase optimized for reliable cDNA synthesis over a wide dynamic range of input RNA. The unique blend of oligo (dT) and random primers in the ReadyScript cDNA Synthesis Mix works exceptionally well with a wide variety of targets. It is optimized for the production of targets 1kb in length. ReadyScript cDNA Synthesis Mix produces excellent results in both real-time and conventional RT-PCR.