Learn More
Sigma Aldrich Fine Chemicals Biosciences ReadyShieldR Protease Inhi
Catalog No. 502904870
Encompass
This protease inhibitor cocktail has been optimized and tested for mammalian cell and tissue extracts. It contains inhibitors with a broad specificity for serine cysteine acid proteases and aminopeptidases. The cocktail has been tested on extracts from various animal tissues such as human placenta and bovine liver and on extracts of A431 CHO and U937 cells.This cocktail is supplied as a ready-to-use SOLUTIONon using a proprietary non-freezing formulation. ReadyShield PIC0002 is a non-freezing formulation that contains the same inhibitors as is the DMSO based cocktail P8340.
Catalog No. 50-290-4870 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PIC00025ML
This protease inhibitor cocktail has been optimized and tested for mammalian cell and tissue extracts. It contains inhibitors with a broad specificity for serine cysteine acid proteases and aminopeptidases. The cocktail has been tested on extracts from various animal tissues such as human placenta and bovine liver and on extracts of A431 CHO and U937 cells.This cocktail is supplied as a ready-to-use SOLUTIONon using a proprietary non-freezing formulation. ReadyShield PIC0002 is a non-freezing formulation that contains the same inhibitors as is the DMSO based cocktail P8340.