Learn More
Research Products International Corp ReadyStrain Cell Straining Kits, 100µm, Yellow, 50 per package
SDP
Catalog No. 501977678
Encompass
ReadyStrain Kits include cell strainer, 50ml tube and cap. Ideal for performing filtration of cell suspensions quickly and conveniently. All in one kit allows straining, processing of sample and storage. Sterile kit is ready to use with a strainer pre-attached to 50ml centrifuge tube. Ideal kit for opening under hood and reducing contamination risk. Supplied pre-assembled in individually wrapped easy to open "tear-strip' bags.
Includes strainer, 50ml tube and cap
Convenient sample processing
Sterile, pre-assembled
Catalog No. 50-197-7678 Supplier Research Products International Corp Supplier No. 401107
ReadyStrain Kits include cell strainer, 50ml tube and cap. Ideal for performing filtration of cell suspensions quickly and conveniently. All in one kit allows straining, processing of sample and storage. Sterile kit is ready to use with a strainer pre-attached to 50ml centrifuge tube. Ideal kit for opening under hood and reducing contamination risk. Supplied pre-assembled in individually wrapped easy to open "tear-strip' bags.
Includes strainer, 50ml tube and cap
Convenient sample processing
Sterile, pre-assembled