Learn More
Bio Rad Laboratories READYSTRIP 100X PH 7-10 BUFFER
Catalog No. 509012744
Encompass
Buffer; Bio-Rad; ReadyStrip; 100x pH 7-10; Anode and pH range clearly printed on each strip; Includes only ampholytes; 1mL
Catalog No. 50-901-2744 Supplier Bio Rad Laboratories Supplier No. 1632093
Buffer; Bio-Rad; ReadyStrip; 100x pH 7-10; Anode and pH range clearly printed on each strip; Includes only ampholytes; 1mL