Learn More
Bio Rad Laboratories READYSTRIP IPG STRIPS
Catalog No. 509013304
Encompass
ReadyStrip IPG strips provide an immobilized pH gradient on a support strip for use in first-dimension isoelectric focusing separations in 2-D electrophoresis. This 11 cm strip is designed to fit on any polyacrylamide gel.
Catalog No. 50-901-3304 Supplier Bio Rad Laboratories Supplier No. 1632018
ReadyStrip IPG strips provide an immobilized pH gradient on a support strip for use in first-dimension isoelectric focusing separations in 2-D electrophoresis. This 11 cm strip is designed to fit on any polyacrylamide gel.