Learn More
McKesson General Medical Abbott i-STAT Chem8+ Cartridge, 9 Tests, Rapid Results in 2 Minutes, 25 Cartridges
Catalog No. NC2087544
Encompass_Preferred
The Abbott i-STAT Chem8+ Cartridge is a single-use reagent cartridge designed for quick metabolic and renal status assessments from arterial or venous whole blood samples. It ensures lab-quality results within approximately 2 minutes, enhancing clinical decision-making across various healthcare settings. This point-of-care solution stands out due to its advanced biosensor technology and compatibility with the i-STAT 1 System, offering a comprehensive panel of 9 tests, including electrolytes and glucose.
This item is not returnable. View return policy
Catalog No. NC2087544 Supplier McKesson General Medical Supplier No. 1166840
The Abbott i-STAT Chem8+ Cartridge is a single-use reagent cartridge designed for quick metabolic and renal status assessments from arterial or venous whole blood samples. It ensures lab-quality results within approximately 2 minutes, enhancing clinical decision-making across various healthcare settings. This point-of-care solution stands out due to its advanced biosensor technology and compatibility with the i-STAT 1 System, offering a comprehensive panel of 9 tests, including electrolytes and glucose.