Learn More
Pickering Laboratories Inc REAG KIT NINHYDRIN 8BTLS/CS
SDP
Catalog No. NC9728701
Encompass_Preferred
TRIONE Two-part Ninhydrin Reagent (12-month* shelf life before mixing), to prepare case of 4 (900 mL/bottle) Post-column Reagent. Amino Acid Analysis.
Catalog No. NC9728701 Supplier Pickering Laboratories Inc Supplier No. T200
TRIONE Two-part Ninhydrin Reagent (12-month* shelf life before mixing), to prepare case of 4 (900 mL/bottle) Post-column Reagent. Amino Acid Analysis.