Learn More
Lifeline Cell Technology REAG MAMCL CRYO LIFELINE BTL
SDP
Catalog No. 50311918
Encompass_Preferred
FrostaLife is a new solution optimized for the cryopreservation of mammalian cells. Simply count your cells, centrifuge them and resuspend the cell pellet in the desired volume of KryoLife and dispense into cryopreservation vials.
Catalog No. 50-311-918 Supplier Lifeline Cell Technology Supplier No. LM0015
FrostaLife is a new solution optimized for the cryopreservation of mammalian cells. Simply count your cells, centrifuge them and resuspend the cell pellet in the desired volume of KryoLife and dispense into cryopreservation vials.