Learn More
CELLTREAT Scientific Products 5mL Reagent Reservoir, Polystyrene, White, Individually Wrapped, Sterile
SDP
Catalog No. NC2545725
Encompass_Preferred
Reagent Reservoirs are designed to assist in repetitive liquid transfer White color enhances solution visibility Angled design promotes funneling of liquid to maximize retrieval Features include pour spouts on the corners, textured gripping areas on the sides, and a nesting design for easy stacking and storage (except 50mL) 5 & 25mL reservoirs accommodate 8-channel pipettes; 50, 55 & 100mL reservoirs accommodate 12-channel pipettes Unique Pyramid Bottom reservoir enables easy, efficient, one-handed recovery using a single-channel pipette Wide design of Pyramid Bottom reservoir reduces risk of contamination compared to pipetting from a tube
Catalog No. NC2545725 Supplier CELLTREAT Scientific Products Supplier No. 229288
Reagent Reservoirs are designed to assist in repetitive liquid transfer White color enhances solution visibility Angled design promotes funneling of liquid to maximize retrieval Features include pour spouts on the corners, textured gripping areas on the sides, and a nesting design for easy stacking and storage (except 50mL) 5 & 25mL reservoirs accommodate 8-channel pipettes; 50, 55 & 100mL reservoirs accommodate 12-channel pipettes Unique Pyramid Bottom reservoir enables easy, efficient, one-handed recovery using a single-channel pipette Wide design of Pyramid Bottom reservoir reduces risk of contamination compared to pipetting from a tube