Learn More
CHEMOMETEC Reagent S100, 5 l.
SDP
Catalog No. NC1715767
Encompass
Reagent S100 is used to treat the sperm cell sample prior to counting the total number of sperm cells in ejaculates or other sperm cells suspensions. Storage: Room temperature
Catalog No. NC1715767 Supplier CHEMOMETEC Supplier No. 9100100
Reagent S100 is used to treat the sperm cell sample prior to counting the total number of sperm cells in ejaculates or other sperm cells suspensions. Storage: Room temperature