Learn More
Cayman Chemical Reagent 1 (R1), 8 mL
SDP
Catalog No. 502978656
Encompass_Preferred
Reagent 1 (R1) has been tested and formulated to work exclusively with Cayman’s Total and Direct Bilirubin Colorimetric Assay Kit (Item No. 701720).
Catalog No. 50-297-8656 Supplier Cayman Chemical Supplier No. 4004948 ML
Reagent 1 (R1) has been tested and formulated to work exclusively with Cayman’s Total and Direct Bilirubin Colorimetric Assay Kit (Item No. 701720).