Learn More
Cayman Chemical Reagent 2R2 2mL
SDP
Catalog No. 502978470
Encompass_Preferred
Reagent 2 (R2) has been tested and formulated to work exclusively with Cayman’s Total and Direct Bilirubin Colorimetric Assay Kit (Item No. 701720).
Catalog No. 50-297-8470 Supplier Cayman Chemical Supplier No. 4004952 ML
Reagent 2 (R2) has been tested and formulated to work exclusively with Cayman’s Total and Direct Bilirubin Colorimetric Assay Kit (Item No. 701720).