Learn More
ELITechGroup Aerospray Hematology Stat Reagent A, Rinse, 500mL
SDP
Catalog No. NC9416394
Encompass
This item has a minimum order quantity of 4 per supplier requirements. Aqueous Eosin Y Stain; Liquid, Orange; 6 pH Approximately; 1 gal.; Hematology Reagent A Rinse; B.P. 100 deg.C; Freezing Point 0 deg.C; Storage: RT away from direct light
Catalog No. NC9416394 Supplier ELITechGroup Supplier No. SS035A
This item has a minimum order quantity of 4 per supplier requirements. Aqueous Eosin Y Stain; Liquid, Orange; 6 pH Approximately; 1 gal.; Hematology Reagent A Rinse; B.P. 100 deg.C; Freezing Point 0 deg.C; Storage: RT away from direct light