Learn More
StatLab Reagent Alcohol 100%
Description
- Ethanol based, anhydrous, and suitable for use in clinical laboratory protocols
- Ideal for processing and staining protocols that reference 100% alcohol, ethanol, or 100% ethanol in Histology and Cytology laboratories
- 55-Gallon Drum
Specifications
Specifications
| Product Type | Dehydrating Reagent |
| Quantity | 55 ga. |
| Form | Solution |