Learn More
IMEB, Inc Reagent Alcohol, 80%, 1 gallon bottle
SDP
Catalog No. NC1897349
Encompass_Preferred
Tek-Select Reagent Alcohol 80%, 1 Gallon. Tek-Select Reagent Alcohol is composed of 90 parts ethyl alcohol, 5 parts methyl alcohol, and 5 parts isopropyl alcohol, v/v blend. Preferred mixture for dehydrating tissue samples prior to embedding and microscope slide staining. Tek-Select Reagent Alcohols are suitable for most histological procedures. 1 Gallon Bottle.
Catalog No. NC1897349 Supplier IMEB, Inc Supplier No. TA11080
Tek-Select Reagent Alcohol 80%, 1 Gallon. Tek-Select Reagent Alcohol is composed of 90 parts ethyl alcohol, 5 parts methyl alcohol, and 5 parts isopropyl alcohol, v/v blend. Preferred mixture for dehydrating tissue samples prior to embedding and microscope slide staining. Tek-Select Reagent Alcohols are suitable for most histological procedures. 1 Gallon Bottle.