Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ricca Chemical Company Reagent Alcohol, ACS Reagent Grade Anhydrous, Absolute, Ricca Chemical
SDP

Catalog No. R659000038 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
3.8 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R659000038 3.8 L Natural Poly Bottle
6590-16 500 mL Natural Poly Bottle
6590-32 1 L Natural Poly Bottle
659025 10 L Cubitainer
6590-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R659000038 Supplier Ricca Chemical Company Supplier No. R65900003.8A
Add to Cart
Add to Cart
  • Reagent Alcohol, ACS Reagent Grade Anhydrous, Absolute

Chemical Identifiers

Molecular Formula C2H6O

Specifications

Melting Point Approximately -114°C
CAS 64-17-5, 67-63-0, 67-56-1
Color Colorless
Quantity 3.8 L
Packaging Natural Poly Bottle
Synonym Ethyl Alcohol
Boiling Point Approximately 76°C
Molecular Weight (g/mol) 46.07
Percent Purity 89.5 to 91.5% (v/v)
Grade ACS
Chemical Name or Material Alcohol, Reagent , Anhyd., Absolute
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.