Learn More
Sigma Aldrich Fine Chemicals Biosciences Reagent Alcohol anhydrous
Catalog No. 502898783
Encompass
Reagent Alcohol is ethanol that is made non-consumable by the addition of denaturants.
Catalog No. 50-289-8783 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2776494X2L
Reagent Alcohol is ethanol that is made non-consumable by the addition of denaturants.