Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol, Solstice

Catalog No. 60048572 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 L
204 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-572 4 L
60-090-446 204 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-572 Supplier Solstice Supplier No. SR090ZA4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, anhydrous, SDA 3A, 90 ±1%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Specifications

CAS 64-17-5, 67-56-1, 67-63-0
CAS Max % 90.00%, 5.00%, 5.00%
Quantity 4 L
Flash Point 15°C.The physical data is that of the main component.
Packaging Metal Drum
Physical Form Liquid, clear
Purity Grade Notes ACS general use solvents
Boiling Point 78.32°C.The physical data is that of the main component.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.