Learn More
Ethos Biosciences Quick III Solution II, 16oz, Ethos
Catalog No. 5000574180
Encompass_Preferred
Quick III Solution II is a staining reagent designed specifically for the Quick III process. Used as the second staining step in the Diff Quik procedure, it contains a basophilic dye that stains nuclei and certain cellular components, providing clear nuclear detail and contrast. The solution delivers consistent color intensity for reliable slide preparation and rapid microscopic evaluation. Ideal for blood smears, semen, fine needle aspirates, and other cytological specimens, it supports efficient, high-throughput diagnostic workflows.
Catalog No. 50-005-74180 Supplier Ethos Biosciences Supplier No. 531916
Quick III Solution II is a staining reagent designed specifically for the Quick III process. Used as the second staining step in the Diff Quik procedure, it contains a basophilic dye that stains nuclei and certain cellular components, providing clear nuclear detail and contrast. The solution delivers consistent color intensity for reliable slide preparation and rapid microscopic evaluation. Ideal for blood smears, semen, fine needle aspirates, and other cytological specimens, it supports efficient, high-throughput diagnostic workflows.