Learn More
BURKLE INC Wide-necked reagent bottle, HDPE, 16.91 oz. (500 ml), with cap
Catalog No. 5000025606
Encompass_Preferred
The transparent wide-neck reagent bottle with tamper-evident screw cap is ideal for storing, packaging, sampling and transporting granular and pasty media.
The bottles are UN approved for solids.
The bottle has a wide neck opening for easy filling and emptying. The tight-fitting screw cap ensures safe storage of chemicals.
The square cross-section of the bottle provides a secure stand. A recessed labeling area makes labeling or marking easy. The wide-neck chemical bottle is the perfect solution for a variety of material handling needs.
Catalog No. 50-000-25606 Supplier BURKLE INC Supplier No. 03420500
The transparent wide-neck reagent bottle with tamper-evident screw cap is ideal for storing, packaging, sampling and transporting granular and pasty media.
The bottles are UN approved for solids.
The bottle has a wide neck opening for easy filling and emptying. The tight-fitting screw cap ensures safe storage of chemicals.
The square cross-section of the bottle provides a secure stand. A recessed labeling area makes labeling or marking easy. The wide-neck chemical bottle is the perfect solution for a variety of material handling needs.