Learn More
BURKLE INC Narrow-necked reagent bottle, HDPE, 3.38 oz. (100 ml), with cap
Catalog No. 5000025822
Encompass_Preferred
The narrow neck reagent bottles are true all-rounders, perfect for storing, packaging, sampling and safely transporting liquids. Made from transparent HDPE, these bottles allow the contents to be seen.
The blue PP tamper evident screw cap with conical seal and tamper evidence ring ensures leak proof storage. The square bottle shape combined with the recessed label area facilitates labeling and organization. The narrow neck allows for precise dispensing and prevents spillage.
These narrow neck bottles are not only practical, but also aesthetically pleasing. Their square shape fits perfectly on any shelf and helps to store liquids safely and efficiently.
Catalog No. 50-000-25822 Supplier BURKLE INC Supplier No. 03400100
The narrow neck reagent bottles are true all-rounders, perfect for storing, packaging, sampling and safely transporting liquids. Made from transparent HDPE, these bottles allow the contents to be seen.
The blue PP tamper evident screw cap with conical seal and tamper evidence ring ensures leak proof storage. The square bottle shape combined with the recessed label area facilitates labeling and organization. The narrow neck allows for precise dispensing and prevents spillage.
These narrow neck bottles are not only practical, but also aesthetically pleasing. Their square shape fits perfectly on any shelf and helps to store liquids safely and efficiently.