Learn More
Foxx Life Sciences Borosil® Amber Light-Blocking Reagent Bottles with Ground Glass Stoppers and Graduations, 100mL, 10/CS
Catalog No. 501720261
Encompass_Preferred
Borosil® Amber Reagent Bottles are perfect for storing light-sensitive media, chemicals, and reagents. Sturdy ground glass necks for long-term durability. Solid flat-head stoppers, white graduations, and capacities, marking spots. Mechanically strong, chemically compatible, thermally resistant. Amber blocks 300nm to 500nm. Avail. from 60mL to 2L. Reagent Bottles comply with EN ISO 4796-2. 60mL and 125mL options are not covered in ISO standards. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0261 Supplier Foxx Life Sciences Supplier No. 1509016
Borosil® Amber Reagent Bottles are perfect for storing light-sensitive media, chemicals, and reagents. Sturdy ground glass necks for long-term durability. Solid flat-head stoppers, white graduations, and capacities, marking spots. Mechanically strong, chemically compatible, thermally resistant. Amber blocks 300nm to 500nm. Avail. from 60mL to 2L. Reagent Bottles comply with EN ISO 4796-2. 60mL and 125mL options are not covered in ISO standards. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.