Learn More
Foxx Life Sciences Borosil Reagent Bottles with Ground Glass Stoppers and Graduations, 1L (1,000mL), 10/CS
Supplier:  Foxx Life Sciences 1500029
Encompass_Preferred
Borosil Reagent Bottles are perfect for storing media, solvents, and other materials. Sturdy ground glass necks for long-term durability. Solid glass flat-head stoppers, easy-to-read graduations and capacities, marking spots for labeling. Mechanically strong, chemically compatible, thermally resistant. Avail. from 60mL to 2L. Borosil Reagent Bottles comply with EN ISO 4796-2. 60mL & 125mL options are not covered in standards. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Low-Expansion Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-154-2801