Learn More
Foxx Life Sciences Borosil® Reagent Square Bottles with PP Screw Cap (GL80), Graduiations and PP Pouring Ring, 1L (1000mL), 10/CS
Catalog No. 501720349
Encompass_Preferred
Borosil® Reagent Square Bottles are perfect for storing media, solvents, and other materials. Bottles comes with PP Screw cap (GL80). A colorless PP pouring ring aloows for drip free emptying, easy-to-read graduations and capacities, marking spots for labeling. Mechanically strong, chemically compatible, thermally resistant. Avail. from 500mL to 5000mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Low-Expansion Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0349 Supplier Foxx Life Sciences Supplier No. 1506029
Borosil® Reagent Square Bottles are perfect for storing media, solvents, and other materials. Bottles comes with PP Screw cap (GL80). A colorless PP pouring ring aloows for drip free emptying, easy-to-read graduations and capacities, marking spots for labeling. Mechanically strong, chemically compatible, thermally resistant. Avail. from 500mL to 5000mL. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Low-Expansion Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.