Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREXPLUS™ Reagent Bottles with Hollow standard taper Stoppers

Catalog No. 028866 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
Stopper Number:
19
24
29
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Stopper Number
02-886-6 1 L 29
02-886-3 125 mL 19
02-886-4 250 mL 19
02-886-5 500 mL 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Do not place over direct heat or flame.
Catalog No. 02-886-6 Supplier Corning™ Supplier No. 615001L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Do not place over direct heat or flame.

Feature special coating for added safety

Features a protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Autoclavable (121°C) and resistant to thermal shock. Bottle has a ground neck with a standard taper stopper.

For stopper only, see Cat. No. 7575.



Warning: Do not place over direct heat or flame. Do not heat above 121°C moist heat or 110°C dry heat.

Not recommended for dry heat sterilization. Will Melt.
TRUSTED_SUSTAINABILITY

Specifications

Capacity 33.81 oz., 1000 mL
Capacity (Metric) 1 L
Coating Pyrexplus
Diameter (English) 4.21 in.
Diameter (English) Outer Neck 1.77 in.
Diameter (Metric) Outer Neck 45 mm
Material Borosilicate Glass
No. per Case 6/Cs.
No. per Pack 1/Pk.
Sterility Non-sterile
Stopper Number 29
Closure Color Orange
Mouth Narrow
Height (Metric) 235 mm
Height (English) 9.25 in.
Closure Material Polypropylene
Diameter (Metric) 107 mm
Closure Type Plug Seal
Capacity (English) 33.81 oz.
Shape Cylindrical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.