Learn More
ELITechGroup Aerospray Hematology Stat Reagent C, Eosin Stain, 500mL
SDP
Catalog No. NC9416396
Encompass
This item has a minimum order quantity of 4 per supplier requirements. Aqueous Eosin Y Stain; Liquid, Red; 1 gal.; Hematology Reagent C Eosin Stain; B.P. 100 deg.C; Freezing Point 0 deg.C; Storage: RT away from direct light
Catalog No. NC9416396 Supplier ELITechGroup Supplier No. SS035C
This item has a minimum order quantity of 4 per supplier requirements. Aqueous Eosin Y Stain; Liquid, Red; 1 gal.; Hematology Reagent C Eosin Stain; B.P. 100 deg.C; Freezing Point 0 deg.C; Storage: RT away from direct light