Learn More
Ethos Biosciences Quick III Solution I, 1gal, Ethos
Catalog No. 5000574363
Encompass_Preferred
Quick III Solution I is a staining reagent designed specifically for the Quick III process. Used as the first staining step following fixation, it contains an eosinophilic dye that stains cytoplasmic components and background material, providing essential contrast for cellular evaluation. The solution delivers consistent color intensity for reliable slide preparation and rapid microscopic results. Ideal for blood smears, semen, fine needle aspirates, and other cytological specimens, it supports efficient, high-throughput diagnostic workflows.
Catalog No. 50-005-74363 Supplier Ethos Biosciences Supplier No. 5329G
Quick III Solution I is a staining reagent designed specifically for the Quick III process. Used as the first staining step following fixation, it contains an eosinophilic dye that stains cytoplasmic components and background material, providing essential contrast for cellular evaluation. The solution delivers consistent color intensity for reliable slide preparation and rapid microscopic results. Ideal for blood smears, semen, fine needle aspirates, and other cytological specimens, it supports efficient, high-throughput diagnostic workflows.