Learn More
CLICKBIO INC Smarter Reagent Fill Station Tubing Set (5ft.)
Catalog No. NC2669861
Encompass
The Tubing Set consists of a fill tube, a drain tube, and clips to attach to the rail on the Deck Module. Case of 5 x 5ft tubing sets.
Catalog No. NC2669861 Supplier CLICKBIO INC Supplier No. CBAFSTS10
The Tubing Set consists of a fill tube, a drain tube, and clips to attach to the rail on the Deck Module. Case of 5 x 5ft tubing sets.