Learn More
Ethos Biosciences Formaldehyde 37%, 5gal, Ethos
Catalog No. 5000574246
Encompass_Preferred
Formaldehyde is a versatile chemical reagent suitable for pathology and general laboratory applications. Concentrated at 37%. Commonly used as a tissue fixative and preservative, it is ideal for specimen preservation, fixation protocols, and reagent preparation. This clear, colorless solution provides consistent performance for reliable tissue processing and microscopic evaluation. It is also widely used in chemical synthesis, cleaning, and general laboratory procedures. Supplied in chemically compatible containers, formaldehyde is appropriate for clinical, research, pharmaceutical, and industrial laboratory environments.
Catalog No. 50-005-74246 Supplier Ethos Biosciences Supplier No. 33235G
Formaldehyde is a versatile chemical reagent suitable for pathology and general laboratory applications. Concentrated at 37%. Commonly used as a tissue fixative and preservative, it is ideal for specimen preservation, fixation protocols, and reagent preparation. This clear, colorless solution provides consistent performance for reliable tissue processing and microscopic evaluation. It is also widely used in chemical synthesis, cleaning, and general laboratory procedures. Supplied in chemically compatible containers, formaldehyde is appropriate for clinical, research, pharmaceutical, and industrial laboratory environments.