Learn More
Bachem Biotinyl-PEG2-maleimide
SDP
Supplier:  Bachem 4095746.0050
Encompass_Preferred
Biotinyl-PEG2-maleimide, 305372-39-8, C23H35N5O7S, Mr 525.63, 50mg. EZ-Link Maleimide-PEG2-Biotin, Biotin-PEG2-Maleimide. Reagent for linking biotin by a short polar and flexible spacer to cysteine.
Catalog No. 50-194-7401