Learn More
Ethos Biosciences New Methylene Blue, 4oz, Ethos
Catalog No. 5000574326
Encompass_Preferred
New Methylene Blue is a staining reagent commonly used for reticulocyte staining, it enables visualization of residual ribonucleic material within immature red blood cells for accurate reticulocyte counts. The solution provides clear contrast and consistent staining for reliable microscopic evaluation. Suitable for routine hematology workflows and research procedures, New Methylene Blue supports dependable cell morphology assessment and analytical results. Supplied in chemically compatible containers, it is appropriate for clinical, research, and educational laboratory environments.
Catalog No. 50-005-74326 Supplier Ethos Biosciences Supplier No. 555104
Add to Cart
Add to Cart
New Methylene Blue is a staining reagent commonly used for reticulocyte staining, it enables visualization of residual ribonucleic material within immature red blood cells for accurate reticulocyte counts. The solution provides clear contrast and consistent staining for reliable microscopic evaluation. Suitable for routine hematology workflows and research procedures, New Methylene Blue supports dependable cell morphology assessment and analytical results. Supplied in chemically compatible containers, it is appropriate for clinical, research, and educational laboratory environments.