Learn More
Ethos Biosciences Giemsa Stain Azure B, 16oz, Ethos
Catalog No. 5000574352
Encompass_Preferred
Giemsa Azure B is a staining reagent designed for hematology, cytology, and microbiology applications. Commonly used as a component of Romanowsky-type stains, it provides clear nuclear and cytoplasmic detail for reliable microscopic evaluation. Ideal for staining peripheral blood smears, bone marrow specimens, and certain microorganisms, it supports accurate cell morphology assessment and differential staining results. The formulation delivers consistent color development for dependable slide preparation in routine and specialized workflows. Supplied in chemically compatible containers, Giemsa Azure B is suitable for clinical, research, and educational laboratory environments.
Catalog No. 50-005-74352 Supplier Ethos Biosciences Supplier No. 554516
Giemsa Azure B is a staining reagent designed for hematology, cytology, and microbiology applications. Commonly used as a component of Romanowsky-type stains, it provides clear nuclear and cytoplasmic detail for reliable microscopic evaluation. Ideal for staining peripheral blood smears, bone marrow specimens, and certain microorganisms, it supports accurate cell morphology assessment and differential staining results. The formulation delivers consistent color development for dependable slide preparation in routine and specialized workflows. Supplied in chemically compatible containers, Giemsa Azure B is suitable for clinical, research, and educational laboratory environments.