Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent for Hydrogen Peroxide, 15 mL, Lovibond™

Catalog No. 361055884 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
36-105-5884 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-105-5884 Supplier Tintometer Group Supplier No. 424991
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to measure hydrogen peroxide in water samples with Lovibond colorimeters.

  • Low range method measuring 1 to 50 mg/L
  • High range method measuring 40 to 500 mg/L

Specifications

pH Range 0 to 14
Volume (Metric) 10 mL
For Use With (Equipment) Colorimeter
Type Hydrogen Peroxide Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.