Learn More
Aqua Solutions Reagent Kit for EZ1016 Total Hardness Analyzer, 2 L Acid, 2 L Buffer, 2 L Color, 2 L EDTA Solutions, 100 mL Calcium Standard Solution, Ready-to-use
SDP
Catalog No. NC2938927
Encompass
Reagent Kit for EZ1016 Total Hardness Analyzer, 2 L Acid, 2 L Buffer, 2 L Color, 2 L EDTA Solutions, 100 mL Calcium Standard Solution, Ready-to-use
Catalog No. NC2938927 Supplier Aqua Solutions Supplier No. HAAPPC1016KTO
Add to Cart
Add to Cart
Reagent Kit for EZ1016 Total Hardness Analyzer, 2 L Acid, 2 L Buffer, 2 L Color, 2 L EDTA Solutions, 100 mL Calcium Standard Solution, Ready-to-use