Learn More
Hach Company Reagent Kit for EZ1031 Phosphate Analyzer (Color and Standard Solutions)
Supplier:  Hach Company APPC1031KTO
Encompass
Reagent kit containing ready-to-use color and standard solutions for phosphate measurement. The kit reduces hands-on preparation and simplifies workflows for routine online phosphate analysis.
  • Ready-to-use color and standard solutions for phosphate measurement.
  • Reduces hands-on reagent preparation and potential for handling errors.
  • Simplifies routine monitoring workflows and decreases maintenance time.
  • Kit includes a color solution and a phosphate standard solution.
  • Intended for use with online phosphate analyzers.
Catalog No. NC2175798