Learn More
Med Vet International HbA1c Test Kit DCA™ Systems HbA1c Test CLIA Waived, 10311134, 10 Tests/Kit
Catalog No. 503044992
Encompass_Preferred
Only 1 µL of whole blood needed for HbA1c testing Results in 6 minutes No sample or reagent preparation needed allowing for simple and efficient operation
Catalog No. 50-304-4992 Supplier Med Vet International Supplier No. 213834KT
Only 1 µL of whole blood needed for HbA1c testing Results in 6 minutes No sample or reagent preparation needed allowing for simple and efficient operation