Learn More
StatLab Reagent Labels

Description
- Reagent Labels are solvent, moisture, scratch, and tear resistant
- Labels are extremely durable and long-lasting
Specifications
Specifications
| Adhesive Type | Peel and Stick |
| Coating | None |
| Color | White |
| Finish | Gloss |
| Format | Label |
| Material | Polyester |
| Printable Sides | Front |
| Product Type | Reagent Label |
| Sterility | Non-sterile |
| Quantity | 1 Each |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.