Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab, Inc. Reagent Labels

Catalog No. 22505478
Change view
Click to view available options
Color:
Blue
Fluorescent Pink
Green
Orange
Yellow
Display Text:
Date Received Label
Not QC'd Label
Parallel Testing Complete Label
Reagent Label
Received Label
Refrigerate Label
Store At Room Temp Label
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Display Text
22-505-478 Fluorescent Pink Parallel Testing Complete Label
22-505-450 Blue Refrigerate Label
22-505-424 Fluorescent Pink Date Received Label
22-505-475 Green Store At Room Temp Label
22-505-474 Yellow Reagent Label
22-505-473 Green Received Label
22-505-437 Orange Not QC'd Label
22-505-436 Green Date Received Label
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22-505-478 Supplier Marketlab, Inc. Supplier No. 6460
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labels designed for clear and convenient identification of reagents.

  • Designed for reagent identification and labeling applications
  • Supports clear and convenient labeling of reagents
  • Supplied in blister packaging for convenient handling and storage

Specifications

Adhesive Type Peel and Stick
Coating None
Color Fluorescent Pink
Finish Opaque
Format Label
Material Paper
Print Technology None
Product Type Label
Sterility Non-sterile
Display Text Parallel Testing Complete Label
Length (English) 0.625 in.
Width (English) 1.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.