Learn More
Waters Corp Reagent Manager, Single Piston, Pulse-Dampened, 2,500 psi
Catalog No. NC2868612 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Reagent Manager is designed for post-column reagent delivery, utilizing a single piston, pulse-dampened pumping system. It minimizes baseline noise and offers precise control for various laboratory applications.
  • Single piston, pulse-dampened pumping system.
  • Minimizes baseline noise via built-in pulse damper with pressure transducer.
  • RoHS compliant.
  • Single-pump; stackable for two-stage applications.
  • Inert polymeric flow path for broad chemical compatibility.
  • Self-flushing piston head.
  • Front panel control with LED display.
  • Automatic pump shutdown at user-programmable pressure limits.
  • Remote stop/start inputs/outputs for integration and safety.
  • Prime/purge valve and keypad for quick solvent changes.
  • Automatic piston wash for improved reliability and seal life.
Catalog No. NC2868612 Supplier Waters Corp Supplier No. 725000556
Add to Cart
Add to Cart
Request a Quote
The Reagent Manager is designed for post-column reagent delivery, utilizing a single piston, pulse-dampened pumping system. It minimizes baseline noise and offers precise control for various laboratory applications.
  • Single piston, pulse-dampened pumping system.
  • Minimizes baseline noise via built-in pulse damper with pressure transducer.
  • RoHS compliant.
  • Single-pump; stackable for two-stage applications.
  • Inert polymeric flow path for broad chemical compatibility.
  • Self-flushing piston head.
  • Front panel control with LED display.
  • Automatic pump shutdown at user-programmable pressure limits.
  • Remote stop/start inputs/outputs for integration and safety.
  • Prime/purge valve and keypad for quick solvent changes.
  • Automatic piston wash for improved reliability and seal life.