Learn More
Waters Corp Waters Corporation Performance Maintenance Kit, Reagent Manager, HPLC, Annual
Catalog No. 50818986 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit contains all the necessary parts for annual maintenance to ensure the Reagent Manager operates at peak performance. It is designed for HPLC systems.
  • Includes piston seal kit
  • Includes seal flush diaphragm
  • Includes two filters
  • Includes two filter elements
  • Includes four check valve capsules
  • Includes two 2µm inline filters
  • Designed for annual replacement
  • One unit per package
Catalog No. 50-818-986 Supplier Waters Corp Supplier No. 201000102
This kit contains all the necessary parts for annual maintenance to ensure the Reagent Manager operates at peak performance. It is designed for HPLC systems.
  • Includes piston seal kit
  • Includes seal flush diaphragm
  • Includes two filters
  • Includes two filter elements
  • Includes four check valve capsules
  • Includes two 2µm inline filters
  • Designed for annual replacement
  • One unit per package