Learn More
Reagent N, Lovibond™
Catalog No. 09050184 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
30 mL
Catalog No. Volume (Metric)
09-050-184 30 mL
1 options
Catalog No. 09-050-184 Supplier Tintometer Group Supplier No. 697271
Add to Cart
Add to Cart
Replacement reagent N for Truog soil test kit.
  • Used with Lovibond Soil Test Kit to measure soil pH from 4.0 to 8.5
  • This item is Reagent N only
Specifications
pH Range 0 to 14
Volume (Metric) 30 mL
Type Soil Test Reagent