Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ Reagent N,N-Dimethylalphanaphthylamine

Catalog No. 01549891 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-549-891 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-549-891 Supplier Beckman Coulter™ Supplier No. B101545
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reagent N,N-Dimethylalphanaphthylamine for use with Beckman Coulter analyzers.

  • For use with Beckman Coulter analyzers
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5%
For Use With (Equipment) Beckman Coulter Analyzers
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.