Learn More
Ethos Biosciences Periodic Acid 0.5%, 16oz, Ethos
Catalog No. 5000574333
Encompass_Preferred
Periodic Acid 0.5% is a oxidizing reagent designed for histology staining applications. Commonly used in Periodic Acid–Schiff (PAS) staining procedures, it oxidizes carbohydrates to form aldehyde groups, enabling accurate visualization of glycogen, mucins, and other polysaccharides. The clear, colorless solution provides consistent and controlled oxidation for reliable staining results and reproducible microscopic evaluation. Suitable for routine and specialized PAS workflows, Periodic Acid 0.5% supports dependable performance in clinical, research, and educational laboratories. Supplied in chemically compatible containers for convenient laboratory use.
Catalog No. 50-005-74333 Supplier Ethos Biosciences Supplier No. 337616
Periodic Acid 0.5% is a oxidizing reagent designed for histology staining applications. Commonly used in Periodic Acid–Schiff (PAS) staining procedures, it oxidizes carbohydrates to form aldehyde groups, enabling accurate visualization of glycogen, mucins, and other polysaccharides. The clear, colorless solution provides consistent and controlled oxidation for reliable staining results and reproducible microscopic evaluation. Suitable for routine and specialized PAS workflows, Periodic Acid 0.5% supports dependable performance in clinical, research, and educational laboratories. Supplied in chemically compatible containers for convenient laboratory use.