Learn More
Lonza Walkersville ReagentPack Subculture Reagents, 100 mL
Catalog No. NC9144838
Encompass
ReagentPack Subculture Kits have been designed specifically for the passaging of primary cell types. Each kit contains 100 mL each of: Trypsin/EDTA, Trypsin Neutralizing Solution, HEPES Buffered Saline.The ReagentPack Subculture Reagents kit is recommended for use with Lonza's primary cells to ensure effective trypsinization and a successful cell passaging process.Content:1 x Bottle Trypsin/EDTA 100 mL1 x Bottle Trypsin Neutralizing Solution 100 mL1 x Bottle HEPES Buffered Saline Solution 100 mL
Catalog No. NC9144838 Supplier Lonza Walkersville Supplier No. CC5034
ReagentPack Subculture Kits have been designed specifically for the passaging of primary cell types. Each kit contains 100 mL each of: Trypsin/EDTA, Trypsin Neutralizing Solution, HEPES Buffered Saline.The ReagentPack Subculture Reagents kit is recommended for use with Lonza's primary cells to ensure effective trypsinization and a successful cell passaging process.Content:1 x Bottle Trypsin/EDTA 100 mL1 x Bottle Trypsin Neutralizing Solution 100 mL1 x Bottle HEPES Buffered Saline Solution 100 mL