Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Reagent Refill for Digital Tap Water Tour
SDPSCI_ED

Catalog No. S44606 Shop All LaMotte Company Products
Change view
Click to view available options
Class Size:
50 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S44606 50 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S44606 Supplier LaMotte™ Supplier No. R597201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement reagents for tap water Tour.

  • Set of consumable reagents as supplied in the original tap water Tour kit

Specifications

Grade K to 12
Includes Replacement Reagents
Class Size 50 Students
Product Type Reagent Refill
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.