Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Reagent Refills
SDPSCI_ED

Catalog No. S72311 Shop All LaMotte Company Products
Change view
Click to view available options
Type:
Alkalinity Reagent B
Ammonia Reagent 1
Chlorine Reagent 1
Chlorine Reagent 3
Chlorine Reagent Refill
DPD 1 Powder
Ferroin Indicator
Mixed Acid
Nitrogen Reducing Reagent
Potassium Iodide
Sulfide 1
Sulfide 2
Sulfide Test 3
Sulfide Test 4
Volume (English):
0.01 lb.
0.5 oz.
1.01 oz.
2.02 oz.
0.22 lb.
16.9 oz.
32.1 oz.
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Volume (English)
S72311 Chlorine Reagent 1 16.9 oz.
S72312 Chlorine Reagent 3 16.9 oz.
S72318 Sulfide 1 2.02 oz.
S72319 Sulfide 2 2.02 oz.
S72322 Ammonia Reagent 1 1.01 oz.
S72310 Alkalinity Reagent B 2.02 oz.
S72393 Nitrogen Reducing Reagent 0.01 lb.
S72384 Chlorine Reagent 1 0.5 oz.
S72386 DPD 1 Powder 0.22 lb.
S72394 Mixed Acid 2.02 oz.
S72321 Chlorine Reagent Refill 16.9 oz.
S72339 Sulfide Test 3 2.02 oz.
S72340 Sulfide Test 4 2.02 oz.
S72344 Ferroin Indicator 16.9 oz.
S72345 Potassium Iodide 32.1 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. S72311 Supplier LaMotte™ Supplier No. 4498L
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Testing supplies for the discerning scientist

Refills for sulfide tests, potassium iodide, chlorine, ferroine indicators and more

Specifications

Volume (Metric) 500 mL
Storage Requirements 25°C
Type Chlorine Reagent 1
Volume (English) 16.9 oz.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.