Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Reagent Refills for Digital Watershed Tour
SDPSCI_ED

Catalog No. S44604 Shop All LaMotte Company Products
Change view
Click to view available options
Class Size:
50 Students (8 Groups)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S44604 50 Students (8 Groups)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S44604 Supplier LaMotte™ Supplier No. R5974
Add to Cart
Add to Cart

Replacement reagents for 5974 Watershed Tour.

  • Keep introducing students to stream and water ecosystems through a classroom-based exploration of the virtual Big River watershed with these replacement reagents

Specifications

Grade K to 12
Includes Replacement Reagents
Class Size 50 Students (8 Groups)
Product Type Reagent Refill
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.