Learn More
Research Products International Corp Reagent Reservoir, Chiller, 100mL
SDP
Catalog No. 501978276
Encompass
Reagent Chiller keeps solution at <5°C for up to two hours. It is custom fit for 100ml Reservoir. The Reagent Chiller contains non-toxic gel refrigerant. It is durable and reusable. Ideal for use as a refill station.
Catalog No. 50-197-8276 Supplier Research Products International Corp Supplier No. 248232
Reagent Chiller keeps solution at <5°C for up to two hours. It is custom fit for 100ml Reservoir. The Reagent Chiller contains non-toxic gel refrigerant. It is durable and reusable. Ideal for use as a refill station.