Learn More
Research Products International Corp Reagent Reservoir Basins, 100ml Capacity, Sterile, Individually Wrapped, 50 per Case
SDP
Catalog No. 501367550
Encompass
Ideal reagent reservoir basins are designed with narrow 'V' bottom for complete sample aspiration. Reagent Basins accept 8 or 12 channel pipettes and feature rigid, durable walls with pour spouts to facilitate spill-free pouring. Unique packaging offers quick and easy open tear strip.
Catalog No. 50-136-7550 Supplier Research Products International Corp Supplier No. 246284
Ideal reagent reservoir basins are designed with narrow 'V' bottom for complete sample aspiration. Reagent Basins accept 8 or 12 channel pipettes and feature rigid, durable walls with pour spouts to facilitate spill-free pouring. Unique packaging offers quick and easy open tear strip.