Learn More
Mascon Medical Reagent Reservoir, Non Sterile, Polystyrene, Drip-free. Designed for use with all 8-channel and 12-channel pipettes, 100ml, 1EA/Bag, 100EA/Case
SDP
Catalog No. NC2180271
Encompass_Preferred
Mascon Medical™ Reagent Reservoirs are perfect for your everyday multi-channel pipetting needs. To ensure simple and efficient operation, our reservoirs are made from virgin Polystyrene with pour-off spouts on each of the 4 corners. The V bottom provides a narrow-tapered trough to minimize reagent loss and maximize recovery. These non sterile 100ml reservoirs are sealed in an easy-to-open tear-strip Polyethylene packaging and are designed for use with all 8-channel and 12-channel pipettes.Specifications: Volume: 100ml Qty. / Case: 100 Material: Polystyrene Color: White Non Sterile: Yes Packaging: Easy-to-open, tear-strip Polyethylene packaging Packaging Format: 1EA/Bag, 100EA/Case
This item is not returnable. View return policy
Catalog No. NC2180271 Supplier Mascon Medical Supplier No. FSRR100
Mascon Medical™ Reagent Reservoirs are perfect for your everyday multi-channel pipetting needs. To ensure simple and efficient operation, our reservoirs are made from virgin Polystyrene with pour-off spouts on each of the 4 corners. The V bottom provides a narrow-tapered trough to minimize reagent loss and maximize recovery. These non sterile 100ml reservoirs are sealed in an easy-to-open tear-strip Polyethylene packaging and are designed for use with all 8-channel and 12-channel pipettes.Specifications: Volume: 100ml Qty. / Case: 100 Material: Polystyrene Color: White Non Sterile: Yes Packaging: Easy-to-open, tear-strip Polyethylene packaging Packaging Format: 1EA/Bag, 100EA/Case